Thermo Scientific Chemicals

Diethyl 1,1-cyclobutanedicarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: B22140.18
50 g, Each
Thermo Scientific Chemicals

Diethyl 1,1-cyclobutanedicarboxylate, 95%, Thermo Scientific Chemicals

Catalog number: B22140.18
50 g, Each
Quantity
Catalog number: B22140.18
also known as B22140-18
Price (USD)
Price: 184.00
Online price: 158.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3779-29-1
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥94.0%
Refractive Index
1.4330-1.4390 @ 20?C

Description

Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl 1,1-cyclobutanedicarboxylate is used as a pharmaceutical intermediate. Used as intermediate in the production of Carboplatin.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text