Thermo Scientific Chemicals

4,4,4-Trifluorobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: B22186.03
1 g, Each
Thermo Scientific Chemicals

4,4,4-Trifluorobutyric acid, 97%, Thermo Scientific Chemicals

Catalog number: B22186.03
1 g, Each
Quantity
Catalog number: B22186.03
also known as B22186-03
Price (USD)
Price: 57.10
Online price: 48.65
Your price:
Quantity
-

Chemical Identifiers

CAS
406-93-9
IUPAC Name
4,4,4-trifluorobutanoate
Molecular Formula
C4H4F3O2
InChI Key
WTUCTMYLCMVYEX-UHFFFAOYSA-M
SMILES
[O-]C(=O)CCC(F)(F)F
Appearance (Color)
White
Form
Crystals or powder or crystalline powder or lumps or fused solid
Assay (GC)
≥96.0%
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Melting Point (clear melt)
27.0-36.0?C

Description

4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4,4-Trifluorobutyric acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text