Thermo Scientific Chemicals

Dichloroacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B22296.22
100 g, Each
Thermo Scientific Chemicals

Dichloroacetyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: B22296.22
100 g, Each
Quantity
Catalog number: B22296.22
also known as B22296-22
Price (USD)
Quantity
-

Chemical Identifiers

CAS
79-36-7
IUPAC Name
2,2-dichloroacetyl chloride
Molecular Formula
C2HCl3O
InChI Key
FBCCMZVIWNDFMO-UHFFFAOYSA-N
SMILES
ClC(Cl)C(Cl)=O
Form
Liquid
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4585-1.4635 @ 20?C
Assay (GC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text