Thermo Scientific Chemicals

Bathocuproin sulfonate disodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B22550.06
5 g, Each
Thermo Scientific Chemicals

Bathocuproin sulfonate disodium salt hydrate, 97%, Thermo Scientific Chemicals

Catalog number: B22550.06
5 g, Each
Quantity
Catalog number: B22550.06
also known as B22550-06
Price (USD)
Price: 419.00
Online price: 356.65
Your price:
Quantity
-

Chemical Identifiers

CAS
52698-84-7
IUPAC Name
disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
Molecular Formula
C26H18N2Na2O6S2
InChI Key
RNGKZLRAVYPLJC-UHFFFAOYSA-L
SMILES
[Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O
Appearance (Color)
White to cream to pale brown
Form
Powder
Assay (HPLC)
≥96.0%
Identification (FTIR)
Conforms
Loss on Drying
≤9%

Description

Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

Solubility
Miscible with water.

Notes
Incompatible with sulfuric acid, sodium metal, potassium metal and calcium carbide.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text