Thermo Scientific Chemicals

Bathocuproin, 98%, Thermo Scientific Chemicals

Catalog number: B22841.06
5 g, Each
Thermo Scientific Chemicals

Bathocuproin, 98%, Thermo Scientific Chemicals

Catalog number: B22841.06
5 g, Each
Quantity
Catalog number: B22841.06
also known as B22841-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4733-39-5
IUPAC Name
2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
Molecular Formula
C26H20N2
InChI Key
STTGYIUESPWXOW-UHFFFAOYSA-N
SMILES
CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1
Appearance (Color)
White to yellow or pale cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5% (non-U.S. sourced material)
Assay from Supplier's CofA
≥97.5% (U.S. sourced material)
Comment
Material sourced in the U.S. and in other countries

Description

It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent for the determination of copper. It acts as an exciton blocking barrier which prohibits excitons diffusion process towards the Al electrode otherwise being quenched. It is the most commonly used buffer layer between acceptor and cathode layer.

Solubility
Soluble in methanol (36mg/100ml), ethanol, 2-propanol, bezene, and acetone. Partly miscible in water.

Notes
Store in cool, dry conditions in a sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text