Thermo Scientific Chemicals

1,2-Dibromopropane, 98%, Thermo Scientific Chemicals

Catalog number: B22994.22
100 g, Each
Thermo Scientific Chemicals

1,2-Dibromopropane, 98%, Thermo Scientific Chemicals

Catalog number: B22994.22
100 g, Each
Quantity
Catalog number: B22994.22
also known as B22994-22
Price (USD)

Chemical Identifiers

CAS
78-75-1
IUPAC Name
1,2-dibromopropane
Molecular Formula
C3H6Br2
InChI Key
XFNJYAKDBJUJAJ-UHFFFAOYNA-N
SMILES
CC(Br)CBr
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5175-1.5215 @ 20?C
Assay (GC)
≥97.5%

Description

1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Dibromopropane was used to study the effect of reductant concentration, reductant contact time and suspension pH on reductive dechlorination of carbon tetrachloride by soil manipulated with Fe(II) and HS-. It was used as internal standard during the determination of nitrogenous disinfection byproduct trichloronitromethane by gas chromatography mass spectrometry.

Solubility
Soluble in water (slightly), ether, and alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text