Thermo Scientific Chemicals

N-Ethylethylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: B23577.06
5 g, Each
Thermo Scientific Chemicals

N-Ethylethylenediamine, 98%, Thermo Scientific Chemicals

Catalog number: B23577.06
5 g, Each
Quantity
Catalog number: B23577.06
also known as B23577-06
Price (USD)
Price: 26.90
Online price: 22.65
Your price:
Quantity
-

Chemical Identifiers

CAS
110-72-5
IUPAC Name
(2-azaniumylethyl)(ethyl)azanium
Molecular Formula
C4H14N2
InChI Key
SCZVXVGZMZRGRU-UHFFFAOYSA-P
SMILES
CC[NH2+]CC[NH3+]
Form
Liquid
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Refractive Index
1.4370-1.4410 @ 20?C
Appearance (Color)
Clear colorless

Description

N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text