Thermo Scientific Chemicals

Methyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B23594.AK
250 mL, Each
Thermo Scientific Chemicals

Methyltrimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B23594.AK
250 mL, Each
Quantity
Catalog number: B23594.AK
also known as B23594-AK
Price (USD)
Price: 43.90
Online price: 37.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1185-55-3
IUPAC Name
trimethoxy(methyl)silane
Molecular Formula
C4H12O3Si
InChI Key
BFXIKLCIZHOAAZ-UHFFFAOYSA-N
SMILES
CO[Si](C)(OC)OC
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.3685-1.3735 @ 20?C

Description

Methyltrimethoxysilane is used as an acid scavenger, for example in the formation of substituted azulenes from allenylsilanes and tropyl­ium tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyltrimethoxysilane is used as an acid scavenger, for example in the formation of substituted azulenes from allenylsilanes and tropyl­ium tetrafluoroborate.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text