Thermo Scientific Chemicals

Di-n-propyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B24056.30
250 g, Each
Thermo Scientific Chemicals

Di-n-propyl phthalate, 98%, Thermo Scientific Chemicals

Catalog number: B24056.30
250 g, Each
Quantity
Catalog number: B24056.30
also known as B24056-30
Price (USD)
Price: 513.00
Online price: 436.65
Your price:
Quantity
-

Chemical Identifiers

CAS
131-16-8
IUPAC Name
1,2-dipropyl benzene-1,2-dicarboxylate
Molecular Formula
C14H18O4
InChI Key
MQHNKCZKNAJROC-UHFFFAOYSA-N
SMILES
CCCOC(=O)C1=CC=CC=C1C(=O)OCCC
Refractive Index
1.4950-1.4990 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-n-propyl phthalate is used to make plasticizers and polymer additives. It is also used in chemical reagents, organic intermediates.

Solubility
Soluble in chloroform, methanol.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text