Thermo Scientific Chemicals

n-Butyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: B24390.AU
1000 mL, Each
Thermo Scientific Chemicals

n-Butyl butyrate, 99%, Thermo Scientific Chemicals

Catalog number: B24390.AU
1000 mL, Each
Quantity
Catalog number: B24390.AU
also known as B24390-AU
Price (USD)
Price: 69.60
Online price: 59.65
Your price:
Quantity
-

Chemical Identifiers

CAS
109-21-7
IUPAC Name
butyl butanoate
Molecular Formula
C8H16O2
InChI Key
XUPYJHCZDLZNFP-UHFFFAOYSA-N
SMILES
CCCCOC(=O)CCC
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.5%
Refractive Index
1.4045-1.4075 @ 20?C

Description

It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is very important for the food and beverages industries and used as perfuming agents. It is a applied as a solvent for resins.

Solubility
Soluble in water. (1 g/L).

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text