Thermo Scientific Chemicals

1,2-Diiodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B24425.03
1 g, Each
Thermo Scientific Chemicals

1,2-Diiodobenzene, 98%, Thermo Scientific Chemicals

Catalog number: B24425.03
1 g, Each
Quantity
Catalog number: B24425.03
also known as B24425-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
615-42-9
Appearance (Color)
Clear, colourless to yellow
Assay (GC)
>97.5%
Refractive Index
> 1.7100
Form
liquid

Description

1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Diiodobenzene is useful for Suzuki reaction. It is also used in the synthesis of purino[8,9-f] phenanthridines. Further, it acts as a precursor for the synthesis of active pharmaceutical ingredients intermediate.

Solubility
Slightly miscible with water.

Notes
Light sensitive and air sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatibile with strong oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text