Thermo Scientific Chemicals

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Catalog number: B24474.14
25 g, Each
Thermo Scientific Chemicals

2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%, Thermo Scientific Chemicals

Catalog number: B24474.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B24474.14
also known as B24474-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
3140-73-6
Assay (unspecified)
≥97.5%
Appearance (Color)
White
Form
Powder

Description

2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

Solubility
Soluble in water, chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text