Thermo Scientific Chemicals

2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: B24503.22
100 g, Each
Thermo Scientific Chemicals

2-Chloropropionyl chloride, 96%, Thermo Scientific Chemicals

Catalog number: B24503.22
100 g, Each
Quantity
Catalog number: B24503.22
also known as B24503-22
Price (USD)
Price: 99.10
Online price: 95.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7623-09-8
IUPAC Name
2-chloropropanoyl chloride
Molecular Formula
C3H4Cl2O
InChI Key
JEQDSBVHLKBEIZ-UHFFFAOYNA-N
SMILES
CC(Cl)C(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥95.0%
Refractive Index
1.4355-1.4420 @ 20?C
Form
Liquid

Description

2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.

Solubility
Reacts with water.

Notes
Store away from oxidizing agents, active metals, bases, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text