Thermo Scientific Chemicals

2-Bromo-5-fluorobenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B24989.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-5-fluorobenzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B24989.06
5 g, Each
Quantity
Catalog number: B24989.06
also known as B24989-06
Price (USD)
Price: 56.10
Online price: 48.65
Your price:
Quantity
-

Chemical Identifiers

CAS
57381-39-2
IUPAC Name
2-bromo-5-fluorobenzonitrile
Molecular Formula
C7H3BrFN
InChI Key
MDHNVHCZDCSTMS-UHFFFAOYSA-N
SMILES
FC1=CC=C(Br)C(=C1)C#N
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (GC)
≥98.0%

Description

2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text