Thermo Scientific Chemicals

Methyl (trimethylsilyl)propiolate, 98%, Thermo Scientific Chemicals

Catalog number: B25245.03
1 g, Each
Thermo Scientific Chemicals

Methyl (trimethylsilyl)propiolate, 98%, Thermo Scientific Chemicals

Catalog number: B25245.03
1 g, Each
Quantity
Catalog number: B25245.03
also known as B25245-03
Price (USD)
Price: 123.00
Online price: 106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
42201-71-8
IUPAC Name
methyl 3-(trimethylsilyl)prop-2-ynoate
Molecular Formula
C7H12O2Si
InChI Key
XXGHAMBKHIDZDC-UHFFFAOYSA-N
SMILES
COC(=O)C#C[Si](C)(C)C
Appearance (Color)
Clear colorless
Refractive Index
1.4380-1.4420 @ 20?C
Assay (GC)
≥97.5%
Form
Liquid

Description

Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text