Thermo Scientific Chemicals

Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25278.09
10 g, Each
Thermo Scientific Chemicals

Methyl 2-hydroxy-5-nitrobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: B25278.09
10 g, Each
Quantity
Catalog number: B25278.09
also known as B25278-09
Price (USD)
Price: 52.60
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17302-46-4
IUPAC Name
2-(methoxycarbonyl)-4-nitrobenzen-1-olate
Molecular Formula
C8H6NO5
InChI Key
UUBFELFUKFJSRD-UHFFFAOYSA-M
SMILES
COC(=O)C1=CC(=CC=C1[O-])[N+]([O-])=O
Appearance (Color)
White to pale cream to cream to orange
Assay (GC)
≥97.5%
Melting Point (clear melt)
113.0-119.0?C
Form
Crystals or powder or crystalline powder

Description

Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text