Thermo Scientific Chemicals

Aniline-2,5-disulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: B25350.14
25 g, Each
Thermo Scientific Chemicals

Aniline-2,5-disulfonic acid, 95%, Thermo Scientific Chemicals

Catalog number: B25350.14
25 g, Each
Quantity
Catalog number: B25350.14
also known as B25350-14
Price (USD)
Price: 160.00
Online price: 139.65
Your price:
Quantity
-

Chemical Identifiers

CAS
98-44-2
IUPAC Name
2-aminobenzene-1,4-disulfonic acid
Molecular Formula
C6H7NO6S2
InChI Key
LDCCBULMAFILCT-UHFFFAOYSA-N
SMILES
NC1=CC(=CC=C1S(O)(=O)=O)S(O)(=O)=O
Appearance (Color)
White to pale cream
Assay (Aqueous acid-base Titration)
≥94.0 to ≤106.0% (dry wt. basis)
Melting Point
>300?C
Water Content (Karl Fischer Titration)
≤2.0%
Form
Powder

Description

Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aniline-2,5-disulfonic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text