Thermo Scientific Chemicals

Methyl 1-cyclohexene-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B25568.14
25 g, Each
Thermo Scientific Chemicals

Methyl 1-cyclohexene-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog number: B25568.14
25 g, Each
Quantity
Catalog number: B25568.14
also known as B25568-14
Price (USD)
Price: 447.00
Online price: 386.65
Your price:
Quantity
-

Chemical Identifiers

CAS
18448-47-0
IUPAC Name
methyl cyclohex-1-ene-1-carboxylate
Molecular Formula
C8H12O2
InChI Key
KXPWRCPEMHIZGU-UHFFFAOYSA-N
SMILES
COC(=O)C1=CCCCC1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4745-1.4795 @20?C

Description

Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and bases. 
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text