Thermo Scientific Chemicals

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: B25572.06
5 g, Each
Thermo Scientific Chemicals

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans, 97%, Thermo Scientific Chemicals

Catalog number: B25572.06
5 g, Each
Quantity
Catalog number: B25572.06
also known as B25572-06
Price (USD)
Price: 67.30
Online price: 57.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17159-80-7
Appearance (Color)
Clear colorless to pale yellow to black
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4635-1.4685 @ 20?C

Description

Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-hydroxycyclohexanecarboxylate, cis + trans is used in the synthesis of other chemicals and as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text