1-(4-Methoxyphenyl)imidazole, 98%
1-(4-Methoxyphenyl)imidazole, 98%
1-(4-Methoxyphenyl)imidazole, 98%
Thermo Scientific Chemicals

1-(4-Methoxyphenyl)imidazole, 98%

CAS: 10040-95-6 | C10H10N2O | 174.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B25621.03
also known as B25621-03
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.50
Save 3.85 (9%)
Each
Chemical Identifiers
CAS10040-95-6
IUPAC Name1-(4-methoxyphenyl)-1H-imidazole
Molecular FormulaC10H10N2O
InChI KeyXNLOIFUGGCCEQX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)N1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)59-67?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only