Thermo Scientific Chemicals

Methyl 2-oxo-2H-pyran-3-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H25932.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2-oxo-2H-pyran-3-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H25932.06
5 g, Each
Quantity
Catalog number: H25932.06
also known as H25932-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
25991-27-9
IUPAC Name
methyl 2-oxo-2H-pyran-3-carboxylate
Molecular Formula
C7H6O4
InChI Key
GJVZWOMUTYNUCE-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=COC1=O
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
70.0-76.0?C
Appearance (Color)
Yellow to orange to brown
Form
Crystals or powder or crystalline powder

Description

Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-oxo-2H-pyran-3-carboxylate is used as a reagent to investigate alfa, beta-unsaturated carbonyl compounds for their cytotoxic profiles against oral human normal and tumor cells.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text