Thermo Scientific Chemicals

O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: H26082.14
25 g, Each
Thermo Scientific Chemicals

O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: H26082.14
25 g, Each
Quantity
Catalog number: H26082.14
also known as H26082-14
Price (USD)
Price: 210.00
Online price: 178.65
Your price:
Quantity
-

Chemical Identifiers

CAS
148893-10-1
IUPAC Name
[(dimethylamino)({3H-[1,2,3]triazolo[4,5-b]pyridin-3-yloxy})methylidene]dimethylazanium; hexafluoro-λ⁵-phosphanuide
Molecular Formula
C10H15F6N6OP
InChI Key
JNWBBCNCSMBKNE-UHFFFAOYSA-N
SMILES
F[P-](F)(F)(F)(F)F.CN(C)C(ON1N=NC2=CC=CN=C12)=[N+](C)C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms

Description

O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate is a coupling reagent and used as an additive in peptide synthesis., It is also involved efficiently to speed up the coupling process and reduces the loss of chiral integrity.

Solubility
Soluble in acetonitrile. Insoluble in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and highly acidic or alkaline materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text