1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%
1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%
1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%
Thermo Scientific Chemicals

1,2,3,4-Tetrahydro-2,2,4,7-tetramethylquinoline, 97%

CAS: 59388-58-8 | C13H19N | 189.302 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Catalog number H26726.AA
also known as H26726-AA
Price (USD)
51.65
Online Exclusive
57.70
Save 6.05 (10%)
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
51.65
Online Exclusive
57.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS59388-58-8
IUPAC Name2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinoline
Molecular FormulaC13H19N
InChI KeyNRWNXIXJZMSDAU-UHFFFAOYNA-N
SMILESCC1CC(C)(C)NC2=CC(C)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to orange to brown
Assay (GC)≥96.0%
Refractive Index1.5450-1.5500 @ 20°C
Identification (FTIR)Conforms
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only