General Description
• Terrific broth is a nutritional medium mainly constituted by peptone, yeast extract, glycerol (as a carbon source), amino acids, and water-soluble vitamins
Application
• Terrific broth is a highly enriched medium used with glycerol to cultivate bacteria, including Escherichia coli and recombinant strains
• Terrific broth medium is commonly used for maintainance of bacterial strains, for cloning, and for protein expression
• It is a certified medium for plasmid and other bacterial cultures in a liquid medium
• A higher cell density is obtained with this media compared to traditional LB broth