Thermo Scientific Chemicals

7-Fluoroisatin, 97%, Thermo Scientific Chemicals

Catalog number: H26827.14
25 g, Each
Thermo Scientific Chemicals

7-Fluoroisatin, 97%, Thermo Scientific Chemicals

Catalog number: H26827.14
25 g, Each
Quantity
Catalog number: H26827.14
also known as H26827-14
Price (USD)
Price: 281.00
Online price: 240.65
Your price:
Quantity
-

Chemical Identifiers

CAS
317-20-4
IUPAC Name
7-fluoro-2,3-dihydro-1H-indole-2,3-dione
Molecular Formula
C8H4FNO2
InChI Key
HGBGVEOXPHGSOS-UHFFFAOYSA-N
SMILES
FC1=CC=CC2=C1NC(=O)C2=O
Appearance (Color)
Red
Assay (HPLC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
190.0-199.0?C
Identification (FTIR)
Conforms

Description

7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Fluoroisatin is used as the intermediate of cardiovascular, anti-inflammatory and bactericidal drugs.

Solubility
Sparingly Soluble in water (0.022 g/L)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text