Thermo Scientific Chemicals

6-Methylpyridine-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H26841.06
5 g, Each
Thermo Scientific Chemicals

6-Methylpyridine-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H26841.06
5 g, Each
Quantity
Catalog number: H26841.06
also known as H26841-06
Price (USD)
Price: 155.00
Online price: 132.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1122-72-1
IUPAC Name
6-methylpyridine-2-carbaldehyde
Molecular Formula
C7H7NO
InChI Key
AHISYUZBWDSPQL-UHFFFAOYSA-N
SMILES
CC1=CC=CC(C=O)=N1
Assay (GC)
≥97.5%
Appearance (Color)
White to brown
Free acid (titration)
≤1.0%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
28.0-34.0°C

Description

6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methyl-2-pyridinecarboxaldehyde was used in the synthesis of 1-(10-cyano- and -10-bromo-5H-dibenzo[a,d]cyclohepten-5-yl)-4-[(arylmethylene)amino]piperazines which are compounds that exhibits anticon vulsant properties. 6-Methylpyridine-2-carboxaldehyde is used to produce 1-(6-methyl-pyridin-2-yl)-hexan-1-one with pent-1-ene at temperature of 100°C. This reaction will need reagent 2-amino-3-picoline and catalysts (Ph3P)3RhCl, Cp2ZrCl2.

Solubility
Soluble in water (750 g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text