Thermo Scientific Chemicals

Pentamethylcyclopentadienylhafnium(IV) trichloride, 98+%, Thermo Scientific Chemicals

Catalog number: H27010.06
5 g, Each
Thermo Scientific Chemicals

Pentamethylcyclopentadienylhafnium(IV) trichloride, 98+%, Thermo Scientific Chemicals

Catalog number: H27010.06
5 g, Each
Quantity
Catalog number: H27010.06
also known as H27010-06
Price (USD)

Chemical Identifiers

CAS
75181-08-7
IUPAC Name
cyclopenta-1,3-dien-1-ide hafnium trichloride pentamethanide
Molecular Formula
C10H20Cl3Hf
InChI Key
DROXWYIBBCYOSR-UHFFFAOYSA-K
SMILES
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[Cl-].[Cl-].[Cl-].[Hf].C1C=CC=[C-]1
Appearance (Color)
White to cream to pale brown or dark purple
Form
Powder
Assay from Suppliers CofA
≥98.0%
Proton NMR
Conforms to structure

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text