Thermo Scientific Chemicals

1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27100.18
50 g, Each
Thermo Scientific Chemicals

1-n-Butyl-4-methylpyridinium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27100.18
50 g, Each
Quantity
Catalog number: H27100.18
also known as H27100-18
Price (USD)
Price: 292.00
Online price: 252.65
Your price:
Quantity
-

Chemical Identifiers

CAS
343952-33-0
IUPAC Name
1-butyl-4-methylpyridin-1-ium; tetrafluoroboranuide
Molecular Formula
C10H16BF4N
InChI Key
VISYYHYJMCAKAF-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCC[N+]1=CC=C(C)C=C1
Appearance (Color)
Brown
Form
Liquid
Comment
Accepted on supplier's Certificate of Analysis
Ion Chromatography
> 98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text