Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27161.18
50 g, Each
Thermo Scientific Chemicals

1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Catalog number: H27161.18
50 g, Each
Quantity
Catalog number: H27161.18
also known as H27161-18
Price (USD)
Price: 260.00
Online price: 248.65
Your price:
Quantity
-

Chemical Identifiers

CAS
244193-52-0
IUPAC Name
3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
Molecular Formula
C12H23BF4N2
InChI Key
GXZCAMSPWNHTAE-UHFFFAOYSA-N
SMILES
F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
Appearance (Color)
Clear colorless to yellow
Form
Viscous liquid
Identification (FTIR)
Conforms
Residual halide
≤100 ppm (by Ion Chromatography)
Ion Chromatography
1-Methyl-3-octylimidazolium : ≥98.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text