Thermo Scientific Chemicals

Methyl 5-methyl-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: H27570.03
1 g, Each
Thermo Scientific Chemicals

Methyl 5-methyl-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: H27570.03
1 g, Each
Quantity
Catalog number: H27570.03
also known as H27570-03
Price (USD)
Price: 62.00
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2527-96-0
IUPAC Name
methyl 5-methylfuran-2-carboxylate
Molecular Formula
C7H8O3
InChI Key
XBYZJUMTKHUJIY-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(C)O1
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4900-1.4950 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

Furans with suitable substituent in the 2- positions may be transformed into other systems with aromatic character with methoxylation of cyclic acetals of 1,4- dicarbonyl compounds, followed by intramolecular condensations in the acid solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furans with suitable substituent in the 2- positions may be transformed into other systems with aromatic character with methoxylation of cyclic acetals of 1,4- dicarbonyl compounds, followed by intramolecular condensations in the ac
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text