Thermo Scientific Chemicals

8-Bromo-1-octanol, 95%, Thermo Scientific Chemicals

Catalog number: H27628.03
1 g, Each
Thermo Scientific Chemicals

8-Bromo-1-octanol, 95%, Thermo Scientific Chemicals

Catalog number: H27628.03
1 g, Each
Quantity
Catalog number: H27628.03
also known as H27628-03
Price (USD)
Price: 54.70
Online price: 51.65
Your price:
Quantity
-

Chemical Identifiers

CAS
50816-19-8
IUPAC Name
8-bromooctan-1-ol
Molecular Formula
C8H17BrO
InChI Key
GMXIEASXPUEOTG-UHFFFAOYSA-N
SMILES
OCCCCCCCCBr
Refractive Index
1.4780-1.4820 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (Silylated GC)
≥94.0%

Description

It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text