Thermo Scientific Chemicals

Dimethylamine, 2M in THF, Thermo Scientific Chemicals

Catalog number: H27665.AU
1000 mL, Each
Thermo Scientific Chemicals

Dimethylamine, 2M in THF, Thermo Scientific Chemicals

Catalog number: H27665.AU
1000 mL, Each
Quantity

Chemical Identifiers

CAS
124-40-3
IUPAC Name
dimethylamine
Molecular Formula
C2H7N
InChI Key
ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES
CNC
Appearance (Color)
Clear colorless to yellow or pale orange
Assay (Aqueous acid-base Titration)
1.95-2.25 M
Form
Liquid
Refractive Index
1.4010-1.4050 @ 20°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylamine is used as a precursor in the preparation of nitrosodimethylamine.

Solubility
Miscible with water and most organic solvents.

Notes
Hygroscopic. Incompatible with oxidizing agents and oxygen,Sodium aluminum hydride, sodium tetrahydroaluminate, lithium tetrahydroaluminate, borane, metal halides, strong acids, caustic alkalis and bromine.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text