Thermo Scientific Chemicals

8-Chloro-1-octanol, 98%, Thermo Scientific Chemicals

Catalog number: H28417.AC
25 mL, Each
Thermo Scientific Chemicals

8-Chloro-1-octanol, 98%, Thermo Scientific Chemicals

Catalog number: H28417.AC
25 mL, Each
Quantity
Catalog number: H28417.AC
also known as H28417-AC
Price (USD)
Price: 133.00
Online price: 113.65
Your price:
Quantity
-

Chemical Identifiers

CAS
23144-52-7
IUPAC Name
8-chlorooctan-1-ol
Molecular Formula
C8H17ClO
InChI Key
YDFAJMDFCCJZSI-UHFFFAOYSA-N
SMILES
OCCCCCCCCCl
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4570-1.4610 @ 20?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text