Thermo Scientific Chemicals

Oxazole-5-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H30179.03
1 g, Each
Thermo Scientific Chemicals

Oxazole-5-carboxylic acid, 98+%, Thermo Scientific Chemicals

Catalog number: H30179.03
1 g, Each
Quantity
Catalog number: H30179.03
also known as H30179-03
Price (USD)
Price: 64.20
Online price: 54.65
Your price:
Quantity
-

Chemical Identifiers

CAS
118994-90-4
IUPAC Name
1,3-oxazole-5-carboxylic acid
Molecular Formula
C4H3NO3
InChI Key
QCGMEWVZBGQOFN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CN=CO1
Appearance (Color)
White to pale cream
Form
Powder
Assay (Aqueous acid-base Titration)
>98.0 to <102.0%
Assay (HPLC)
>98.0%
Chloride content
(as NaCl) < 1.0% by titration

Description

Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Decarboxylative cross-coupling of thiazole and oxazole-5-carboxylic acids with aryl halides is reported.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text