Thermo Scientific Chemicals

N-Boc-N-methyl-L-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H31317.06
5 g, Each
Thermo Scientific Chemicals

N-Boc-N-methyl-L-alanine, 98%, Thermo Scientific Chemicals

Catalog number: H31317.06
5 g, Each
Quantity
Catalog number: H31317.06
also known as H31317-06
Price (USD)
Price: 257.00
Online price: 222.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16948-16-6
IUPAC Name
(2S)-2-{[(tert-butoxy)carbonyl](methyl)amino}propanoic acid
Molecular Formula
C9H17NO4
InChI Key
VLHQXRIIQSTJCQ-LURJTMIESA-N
SMILES
C[C@H](N(C)C(=O)OC(C)(C)C)C(O)=O
Form
Powder
Melting Point (clear melt)
88.0-94.0?C
Appearance (Color)
White
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Optical Rotation
-30.4 ± 0.3? (C=1 in ethanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text