4-Iodo-1-phenyl-1H-pyrazole, 95%
4-Iodo-1-phenyl-1H-pyrazole, 95%
4-Iodo-1-phenyl-1H-pyrazole, 95%
Thermo Scientific Chemicals

4-Iodo-1-phenyl-1H-pyrazole, 95%

CAS: 23889-85-2 | C9H7IN2 | 270.073 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H32612.03
also known as H32612-03
Price (USD)
189.65
Special offer
Online exclusive
Ends: 30-Jun-2026
223.00
Save 33.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
189.65
Special offer
Online exclusive
Ends: 30-Jun-2026
223.00
Save 33.35 (15%)
Each
Chemical Identifiers
CAS23889-85-2
IUPAC Name4-iodo-1-phenyl-1H-pyrazole
Molecular FormulaC9H7IN2
InChI KeyPYJVDCYRDQUUKO-UHFFFAOYSA-N
SMILESIC1=CN(N=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or granular solid
Appearance (Color)White to pale cream
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only