Thermo Scientific Chemicals

5-Chloro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H32912.06
5 g, Each
Thermo Scientific Chemicals

5-Chloro-2-methylbenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog number: H32912.06
5 g, Each
Quantity
Catalog number: H32912.06
also known as H32912-06
Price (USD)
Price: 315.00
Online price: 271.65
Your price:
Quantity
-

Chemical Identifiers

CAS
34981-38-9
IUPAC Name
5-chloro-2-methylbenzene-1-sulfonyl chloride
Molecular Formula
C7H6Cl2O2S
InChI Key
VHBFBNCERXCECQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(Cl)C=C1S(Cl)(=O)=O
Form
Liquid
Refractive Index
1.5675-1.5725 @ 20?C
Assay (GC)
≥96.0%
Appearance (Color)
Clear colorless to yellow to red

Description

5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methylbenzenesulfonyl chloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text