Thermo Scientific Chemicals

1-Boc-4-(2-chloroethyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog number: H33349.06
5 g, Each
Thermo Scientific Chemicals

1-Boc-4-(2-chloroethyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog number: H33349.06
5 g, Each
Quantity
Catalog number: H33349.06
also known as H33349-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
208167-83-3
IUPAC Name
tert-butyl 4-(2-chloroethyl)piperazine-1-carboxylate
Molecular Formula
C11H21ClN2O2
InChI Key
MYOWELLYEZMECA-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCN(CCCl)CC1
Appearance (Color)
Cream to brown or pale yellow
Form
Powder or crystalline powder
Assay (GC)
≥96.0%
Melting Point (clear melt)
59.0-69.0°C

Description

By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By the direct condensation of chloroalkyl piperazine, nitrogen mustard benzaldehyde, and pyrrole anticancer activity to bel-7404 liver cancer cells was found.

Solubility
Slightly Soluble (7.2 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text