Thermo Scientific Chemicals

2-Bromo-6-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog number: H34478.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-6-iodopyridine, 97%, Thermo Scientific Chemicals

Catalog number: H34478.06
5 g, Each
Quantity
Catalog number: H34478.06
also known as H34478-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
234111-08-1
Appearance (Color)
White to cream to brown
Assay (GC)
≥96.0%
Melting Point (clear melt)
137.0-147.0?C
Form
Crystals or powder or crystalline powder

Description

6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text