1,3-Di-tert-butyl-2-methoxybenzene, 99%
1,3-Di-tert-butyl-2-methoxybenzene, 99%
1,3-Di-tert-butyl-2-methoxybenzene, 99%
Thermo Scientific Chemicals

1,3-Di-tert-butyl-2-methoxybenzene, 99%

CAS: 1516-95-6 | C15H24O | 220.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number H37487.18
also known as H37487-18
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
138.65
Online Exclusive
154.00
Save 15.35 (10%)
Each
Chemical Identifiers
CAS1516-95-6
IUPAC Name1,3-di-tert-butyl-2-methoxybenzene
Molecular FormulaC15H24O
InChI KeyOGIJRUWUWSZKAV-UHFFFAOYSA-N
SMILESCOC1=C(C=CC=C1C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow
Assay (GC)≥98.5%
Refractive Index1.5025-1.5055 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only