Thermo Scientific Chemicals

1,3,5-Tris(4-bromophenyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: H37636.06
5 g, Each
Thermo Scientific Chemicals

1,3,5-Tris(4-bromophenyl)benzene, 97%, Thermo Scientific Chemicals

Catalog number: H37636.06
5 g, Each
Quantity
Catalog number: H37636.06
also known as H37636-06
Price (USD)
Price: 208.00
Online price: 178.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7511-49-1
IUPAC Name
4'-bromo-3,5-bis(4-bromophenyl)-1,1'-biphenyl
Molecular Formula
C24H15Br3
InChI Key
HJQRITCAXSBOPC-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(Br)C=C1)C1=CC=C(Br)C=C1
Comment
May contain small amounts of colored product from manufacturing process
Form
Powder or crystalline powder
Appearance (Color)
White to yellow, pale brown or pale pink
Purity (DSC)
≥96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text