Thermo Scientific Chemicals

2-Chloropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: H50099.06
5 g, Each
Thermo Scientific Chemicals

2-Chloropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Catalog number: H50099.06
5 g, Each
Quantity
Catalog number: H50099.06
also known as H50099-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
100704-10-7
IUPAC Name
(2-chloropyridin-4-yl)methanol
Molecular Formula
C6H6ClNO
InChI Key
UDDVPFLXGOBESH-UHFFFAOYSA-N
SMILES
OCC1=CC=NC(Cl)=C1
Appearance (Color)
Off-white to pale yellow
Form
Solid
Proton NMR
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text