Thermo Scientific Chemicals

2-Hydrazinoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H50700.03
1 g, Each
Thermo Scientific Chemicals

2-Hydrazinoquinoline, 97%, Thermo Scientific Chemicals

Catalog number: H50700.03
1 g, Each
Quantity
Catalog number: H50700.03
also known as H50700-03
Price (USD)
Price: 245.00
Online price: 208.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15793-77-8
IUPAC Name
2-hydrazinylquinoline
Molecular Formula
C9H9N3
InChI Key
QMVCLSHKMIGEFN-UHFFFAOYSA-N
SMILES
NNC1=CC=C2C=CC=CC2=N1
Form
Powder or crystalline powder
Assay (GC)
≥96.0% [U.K. Specification]
Comment
Material Purchased in U.K. and U.S.
Appearance (Color)
White to cream or pale yellow to orange to red or brown
Assay from Supplier's CofA
≥96.0% (HPLC) [U.S. Specification]

Description

2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydrazinoquinoline acts as as a derivatization agent for LC-MS-based metabolomic investigation of diabetic ketoacidosis. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text