Thermo Scientific Chemicals

2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H51875.03
1 g, Each
Thermo Scientific Chemicals

2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Catalog number: H51875.03
1 g, Each
Quantity
Catalog number: H51875.03
also known as H51875-03
Price (USD)
Quantity
-

Description

It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text