Thermo Scientific Chemicals

4-Benzyloxy-3-methylbenzeneboronic acid, 96%, Thermo Scientific Chemicals

Catalog number: H53073.06
5 g, Each
Thermo Scientific Chemicals

4-Benzyloxy-3-methylbenzeneboronic acid, 96%, Thermo Scientific Chemicals

Catalog number: H53073.06
5 g, Each
Quantity
Catalog number: H53073.06
also known as H53073-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
338454-30-1
IUPAC Name
[4-(benzyloxy)-3-methylphenyl]boronic acid
Molecular Formula
C14H15BO3
InChI Key
QPXFEWQLALNXEZ-UHFFFAOYSA-N
SMILES
CC1=C(OCC2=CC=CC=C2)C=CC(=C1)B(O)O
Assay (HPLC)
95.0% min

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text