Thermo Scientific Chemicals

1-Pentadecene, 97%, Thermo Scientific Chemicals

Catalog number: H53516.06
5 g, Each
Thermo Scientific Chemicals

1-Pentadecene, 97%, Thermo Scientific Chemicals

Catalog number: H53516.06
5 g, Each
Quantity
Catalog number: H53516.06
also known as H53516-06
Price (USD)
Price: 58.80
Online price: 55.65
Your price:
Quantity
-

Chemical Identifiers

CAS
13360-61-7
Appearance (Color)
Colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥96.0%
Identification (FTIR)
Conforms

Description

1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecene serves as a growth medium for the strain CV2803T. It is a common semiochemical of flour beetles.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text