Thermo Scientific Chemicals

1,3-Difluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H55694.03
1 g, Each
Thermo Scientific Chemicals

1,3-Difluoro-2-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H55694.03
1 g, Each
Quantity
Catalog number: H55694.03
also known as H55694-03
Price (USD)
Price: 74.50
Online price: 64.65
Your price:
Quantity
-

Chemical Identifiers

CAS
19064-24-5
IUPAC Name
1,3-difluoro-2-nitrobenzene
Molecular Formula
C6H3F2NO2
InChI Key
SSNCMIDZGFCTST-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=C(F)C=CC=C1F
Assay (GC)
>97.5%
Appearance (Color)
Clear yellow
Refractive Index
1.4319 @20?C
Form
Liquid

Description

1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoro-2-nitrobenzene, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text