Thermo Scientific Chemicals

3-Ethynylthiophene, 96%, Thermo Scientific Chemicals

Catalog number: H55898.06
5 g, Each
Thermo Scientific Chemicals

3-Ethynylthiophene, 96%, Thermo Scientific Chemicals

Catalog number: H55898.06
5 g, Each
Quantity
Catalog number: H55898.06
also known as H55898-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
67237-53-0
IUPAC Name
3-ethynylthiophene
Molecular Formula
C6H4S
InChI Key
MJHLPKWONJUCFK-UHFFFAOYSA-N
SMILES
C#CC1=CSC=C1
Assay (GC)
96%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Avoid strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text