Thermo Scientific Chemicals

MES hydrate, 99+%

Catalog number: H56472.30
250 g, Each
Thermo Scientific Chemicals

MES hydrate, 99+%

Catalog number: H56472.30
250 g, Each
Quantity
Catalog number: H56472.30
Price (USD)
Price: 148.00
Online price: 125.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1266615-59-1
IUPAC Name
2-(morpholin-4-yl)ethane-1-sulfonic acid
Molecular Formula
C6H13NO4S
InChI Key
SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES
OS(=O)(=O)CCN1CCOCC1
Form
Powder or crystals or crystalline powder
Loss on Drying
≤9.0%
Assay from Supplier's CofA
≥99.0%
Appearance (Color)
White

Description

MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES hydrate is a zwitterionic N-substituted aminosulfonic acid, known as Good’s buffer, is utilized for many biological applications. It is a useful resolving agent for very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Soluble in water. Slightly soluble in organic solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text