Thermo Scientific Chemicals

Benzyl 2-bromoethyl ether, 97%, Thermo Scientific Chemicals

Catalog number: H56872.06
5 g, Each
Thermo Scientific Chemicals

Benzyl 2-bromoethyl ether, 97%, Thermo Scientific Chemicals

Catalog number: H56872.06
5 g, Each
Quantity
Catalog number: H56872.06
also known as H56872-06
Price (USD)
Price: 119.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1462-37-9
IUPAC Name
[(2-bromoethoxy)methyl]benzene
Molecular Formula
C9H11BrO
InChI Key
FWOHDAGPWDEWIB-UHFFFAOYSA-N
SMILES
BrCCOCC1=CC=CC=C1
Assay (GC)
96.0% min

Description

Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text